Assay Details
Target Gene Details
Entrez Gene ID: | 18755 |
Gene Name: | protein kinase C, eta |
Gene Aliases: |
Pkch |
Location: |
Chr.12:73585041-73778185 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkch | NM_008856.3 | 14 | 2452 | NP_032882.2 |
| AK145817.1 | 14 | 2698 | BAE26670.1 | |
| AK155951.1 | 15 | 2795 | BAE33517.1 | |
| AK164044.1 | 14 | 2651 | BAE37603.1 | |
| BC031121.1 | 14 | 2452 | AAH31121.1 |
More Information
Set Membership: |
|

Genomic Map