Assay Details
Target Gene Details
Entrez Gene ID: | 20474 |
Gene Name: | sine oculis-related homeobox 4 |
Gene Aliases: |
AI047561, AREC3, TrexBF |
Location: |
Chr.12:73099609-73114774 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Six4 | NM_011382.2 | 3 | 3592 | NP_035512.1 |
| D50416.1 | 3 | 3592 | BAA08915.1 | |
| D50418.1 | 5 | 3365 | BAA08917.1 |
More Information
Set Membership: |
|

Genomic Map