Assay Details
Target Gene Details
Entrez Gene ID: | 20476 |
Gene Name: | sine oculis-related homeobox 6 |
Gene Aliases: |
Optx2, Six9 |
Location: |
Chr.12:72939945-72944899 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Six6 | NM_011384.4 | 2 | 1777 | NP_035514.1 |
| AK029309.1 | 2 | 1778 | BAC26387.1 | |
| BC138838.1 | 2 | 1750 | ||
| BC138839.1 | 2 | 1750 |
More Information
Set Membership: |
|

Genomic Map