Assay Details
Target Gene Details
Entrez Gene ID: | 20493 |
Gene Name: | solute carrier family 10 (sodium/bile acid cotransporter family), member 1 |
Gene Aliases: |
Ntcp |
Location: |
Chr.12:80953185-80968702 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc10a1 | NM_001177561.1 | NP_001171032.1 | ||
| NM_011387.2 | 4 | 1204 | NP_035517.1 | |
| AB003303.1 | BAA19846.1 | |||
| BC021154.1 | AAH21154.1 | |||
| BC094023.1 | AAH94023.1 | |||
| U95131.1 | AAB81023.1 | |||
| U95132.1 | 4 | 1109 | AAB81024.1 |
More Information
Set Membership: |
|

Genomic Map