Assay Details
Target Gene Details
Entrez Gene ID: | 16323 |
Gene Name: | inhibin beta-A |
Gene Aliases: |
- |
Location: |
Chr.13:16007745-16027723 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inhba | NM_008380.1 | 2 | 467 | NP_032406.1 |
| AK134991.1 | 4 | 918 | BAE22374.1 | |
| AK135474.1 | 1 | 299 | BAE22545.1 | |
| BC053527.1 | 2 | 459 | AAH53527.1 | |
| CT010380.1 | 1 | 295 | CAJ18587.1 | |
| X69619.1 | 2 | 467 | CAA49325.1 |
More Information
Set Membership: |
|

Genomic Map