Assay Details
Target Gene Details
Entrez Gene ID: | 16323 |
Gene Name: | inhibin beta-A |
Gene Aliases: |
- |
Location: |
Chr.13:16007745-16027723 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inhba | NM_008380.1 | 3 | 726 | NP_032406.1 |
| AK134991.1 | 5 | 1177 | BAE22374.1 | |
| AK135474.1 | 2 | 558 | BAE22545.1 | |
| BC053527.1 | 3 | 719 | AAH53527.1 | |
| CT010380.1 | 2 | 554 | CAJ18587.1 | |
| X69619.1 | 3 | 726 | CAA49325.1 |
More Information
Set Membership: |
|

Genomic Map