Assay Details
Target Gene Details
Entrez Gene ID: | 68346 |
Gene Name: | sirtuin 5 |
Gene Aliases: |
0610012J09Rik, 1500032M05Rik, AV001953 |
Location: |
Chr.13:43365439-43395203 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirt5 | NM_178848.3 | 8 | 978 | NP_849179.1 |
| AK002609.1 | 9 | 1007 | ||
| AK005346.1 | 8 | 991 | ||
| BC031770.1 | 8 | 978 | AAH31770.1 |

Genomic Map