Assay Details
Target Gene Details
Entrez Gene ID: | 105355 |
Gene Name: | solute carrier family 17 (sodium phosphate), member 3 |
Gene Aliases: |
AW261723, Npt4 |
Location: |
Chr.13:23836918-23860718 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a3 | NM_001164743.1 | 12 | 2915 | NP_001158215.1 |
| NM_134069.3 | 13 | 3039 | NP_598830.2 | |
| AK143896.1 | 13 | 3011 | ||
| BC034183.1 | 13 | 2844 | AAH34183.1 |
More Information
Set Membership: |
|

Genomic Map