Assay Details
Target Gene Details
Entrez Gene ID: | 74427 |
Gene Name: | ELL associated factor 1 |
Gene Aliases: |
4933403C17Rik, AI666536 |
Location: |
Chr.14:31495079-31509858 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eaf1 | NM_028932.4 | 1 | 196 | NP_083208.1 |
| AK036661.1 | 1 | 167 | ||
| AK044181.1 | 1 | 197 | BAC31806.1 | |
| AK044737.1 | 1 | 133 | BAC32058.1 | |
| BC079658.1 | 1 | 173 | AAH79658.1 |

Genomic Map