Assay Details
Target Gene Details
Entrez Gene ID: | 12677 |
Gene Name: | visual system homeobox 2 |
Gene Aliases: |
Chx10, Hox-10, Hox10, or |
Location: |
Chr.12:84569828-84595457 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsx2 | NM_001301427.1 | 1 | 407 | NP_001288356.1 |
| NM_007701.3 | 1 | 407 | NP_031727.1 | |
| AK143461.1 | 1 | 407 | BAE25386.1 | |
| BC031869.1 | 1 | 383 | AAH31869.1 | |
| BC051245.1 | 1 | 372 | AAH51245.2 | |
| BC058806.1 | 1 | 395 | AAH58806.1 | |
| L34808.1 | 1 | 292 | AAA60431.1 |
More Information
Set Membership: |
|

Genomic Map