Assay Details
Target Gene Details
Entrez Gene ID: | 73828 |
Gene Name: | DDB1 and CUL4 associated factor 4 |
Gene Aliases: |
1110018E21Rik, Wdr21 |
Location: |
Chr.12:83520457-83541994 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf4 | NM_001165256.1 | 13 | 1487 | NP_001158728.1 |
| NM_030246.2 | 14 | 1544 | NP_084522.1 | |
| BC003284.1 | 13 | 1453 | AAH03284.1 | |
| CK330131.1 | 1 | 465 |

Genomic Map