Assay Details
Target Gene Details
Entrez Gene ID: | 320234 |
Gene Name: | coiled-coil domain containing 66 |
Gene Aliases: |
E230015L20Rik |
Location: |
Chr.14:27481090-27508506 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc66 | NM_177111.3 | 14 | 2095 | NP_796085.3 |
| AK087583.1 | 6 | 816 | BAC39936.1 | |
| AK158666.1 | 12 | 1613 | BAE34603.1 | |
| AK170006.1 | 12 | 1562 | ||
| BC070471.1 | 14 | 2095 | ||
| BC072583.1 | 6 | 1212 | AAH72583.1 |

Genomic Map