Assay Details
Target Gene Details
Entrez Gene ID: | 320234 |
Gene Name: | coiled-coil domain containing 66 |
Gene Aliases: |
E230015L20Rik |
Location: |
Chr.14:27481090-27508506 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc66 | NM_177111.3 | 4 | 498 | NP_796085.3 |
| AK145297.1 | 3 | 511 | BAE26352.1 | |
| AK158666.1 | BAE34603.1 | |||
| AK170006.1 | 4 | 546 | ||
| BC070471.1 | 4 | 498 |

Genomic Map