Assay Details
Target Gene Details
Entrez Gene ID: | 239083 |
Gene Name: | cyclin B1 interacting protein 1 |
Gene Aliases: |
Gm288, Hei10, mei4 |
Location: |
Chr.14:50789249-50795983 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnb1ip1 | NM_001111119.1 | 3 | 468 | NP_001104589.1 |
More Information
Set Membership: |
|

Genomic Map