Assay Details
Target Gene Details
Entrez Gene ID: | 494124 |
Gene Name: | calmodulin 5 |
Gene Aliases: |
AY314008, Scarf2 |
Location: |
Chr.13:3854173-3854761 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calm5 | NM_001008706.2 | 2 | 472 | NP_001008706.1 |
| BC138691.1 | 1 | 512 |
More Information
Set Membership: |
|

Genomic Map