Assay Details
Target Gene Details
Entrez Gene ID: | 110616 |
Gene Name: | ataxin 3 |
Gene Aliases: |
2210008M02Rik, AI463012, AI647473, ATX3, MJD1, Mjd, Sca3, ataxin-3 |
Location: |
Chr.12:101918901-101958800 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atxn3 | NM_029705.3 | 11 | 1677 | NP_083981.2 |
| AK030842.1 | 11 | 1617 | BAC27155.1 |
More Information
Set Membership: |
|

Genomic Map