Assay Details
Target Gene Details
Entrez Gene ID: | 319670 |
Gene Name: | echinoderm microtubule associated protein like 5 |
Gene Aliases: |
BC027154, C130068M19Rik |
Location: |
Chr.12:98786604-98901494 on Build GRCm38 |
Assay Gene Location: | Within Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eml5 | NM_001081191.1 | 44 | 7615 | NP_001074660.1 |
| AK044024.1 | 8 | 2268 | BAC31743.1 | |
| AK082287.1 | 1 | 436 | BAC38455.1 | |
| BC027154.1 | AAH27154.1 | |||
| BC158137.1 | 42 | 7087 |

Genomic Map