Assay Details
Target Gene Details
Entrez Gene ID: | 12499 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 5 |
Gene Aliases: |
AI196558, AI987697, Cd39l4, ER-UDPase, NTPDase-5, NTPDase5, Pcph, mNTPase |
Location: |
Chr.12:84373875-84409029 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Entpd5 | NM_001026214.2 | 16 | 2056 | NP_001021385.1 |
| NM_001286049.1 | 15 | 2031 | NP_001272978.1 | |
| NM_001286058.1 | 14 | 1958 | NP_001272987.1 | |
| NM_007647.3 | 15 | 1983 | NP_031673.2 | |
| AF006482.1 | 15 | 1992 | AAC05181.1 | |
| AK002618.1 | 15 | 1956 | BAB22234.1 | |
| AK031581.1 | 16 | 2036 | BAC27461.1 | |
| AK036641.1 | 16 | 2033 | BAC29515.1 | |
| AK045828.1 | 15 | 1969 | BAC32507.1 | |
| AK080265.1 | 16 | 2043 | BAC37862.1 | |
| AK081435.1 | 16 | 2021 | BAC38219.1 | |
| AK088455.1 | 15 | 2012 | BAC40362.1 | |
| AK160950.1 | 13 | 1877 | BAE36110.1 | |
| AK163688.1 | 15 | 1957 | BAE37457.1 | |
| BC015247.1 | 14 | 1941 | AAH15247.1 |
More Information
Set Membership: |
|

Genomic Map