Assay Details
Target Gene Details
Entrez Gene ID: | 268534 |
Gene Name: | syntrophin, gamma 2 |
Gene Aliases: |
2210008K22Rik, 9530013L23, BB121248 |
Location: |
Chr.12:30174480-30373392 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sntg2 | NM_172951.3 | 17 | 1681 | NP_766539.2 |
| AF367760.1 | 17 | 1536 | AAK53400.1 | |
| AK008673.1 | 17 | 1591 | ||
| AK035310.1 | 17 | 1635 | BAC29026.1 | |
| AK087560.1 | 17 | 1620 | ||
| AK137345.1 | 17 | 1641 | ||
| BC138956.1 | 17 | 1595 |

Genomic Map