Assay Details
Target Gene Details
Entrez Gene ID: | 66354 |
Gene Name: | SNW domain containing 1 |
Gene Aliases: |
2310008B08Rik, AW048543, NCoA-62, SKIP, Skiip |
Location: |
Chr.12:87449910-87472299 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snw1 | NM_025507.2 | 14 | 1988 | NP_079783.2 |
| AK009218.1 | 9 | 1343 | BAB26144.1 | |
| AX775977.1 | 15 | 1663 | CAE11688.1 | |
| BC049245.1 | 2 | 665 | AAH49245.1 |
More Information
Set Membership: |
|

Genomic Map