Assay Details
Target Gene Details
Entrez Gene ID: | 16468 |
Gene Name: | jumonji, AT rich interactive domain 2 |
Gene Aliases: |
Jmj, jumonji |
Location: |
Chr.13:44729181-44921643 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jarid2 | NM_001205043.1 | 18 | 4939 | NP_001191972.1 |
| NM_001205044.1 | 19 | 4612 | NP_001191973.1 | |
| NM_021878.3 | 19 | 4658 | NP_068678.1 | |
| AK147226.1 | 18 | 4939 | BAE27780.1 | |
| BC052444.1 | 19 | 4529 | AAH52444.1 | |
| BC060695.1 | 19 | 4529 | AAH60695.1 |
More Information
Set Membership: |
|

Genomic Map