Assay Details
Target Gene Details
Entrez Gene ID: | 57748 |
Gene Name: | junction-mediating and regulatory protein |
Gene Aliases: |
- |
Location: |
Chr.13:93430097-93499808 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmy | NM_021310.3 | 10 | 3363 | NP_067285.2 |
| AB221690.1 | 10 | 2861 | ||
| AF201390.1 | 10 | 2987 | AAF17555.1 | |
| AK035559.1 | 1 | 44 | BAC29105.1 | |
| BC090835.1 | 10 | 3244 | AAH90835.1 |

Genomic Map