Assay Details
Target Gene Details
Entrez Gene ID: | 75477 |
Gene Name: | profilin 3 |
Gene Aliases: |
1700012P12Rik |
Location: |
Chr.13:55414688-55415232 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfn3 | NM_029303.2 | 1 | 457 | NP_083579.1 |
| AK005930.1 | 1 | 449 | BAB24322.1 | |
| BC048629.1 | 1 | 457 | AAH48629.1 |
More Information
Set Membership: |
|

Genomic Map