Assay Details
Target Gene Details
Entrez Gene ID: | 67881 |
Gene Name: | magnesium-dependent phosphatase 1 |
Gene Aliases: |
1810034K20Rik, AI035604 |
Location: |
Chr.14:55657879-55660508 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdp1 | NM_023397.4 | 6 | 1318 | NP_075886.1 |
| NR_028316.1 | 5 | 1387 | ||
| AK007694.1 | 5 | 1331 | BAB25194.1 | |
| BC046613.1 | 6 | 1241 | AAH46613.1 | |
| CR519902.1 |
More Information
Set Membership: |
|

Genomic Map