Assay Details
Target Gene Details
Entrez Gene ID: | 13983 |
Gene Name: | estrogen receptor 2 (beta) |
Gene Aliases: |
ER[b], ERbeta, Estrb |
Location: |
Chr.12:76120419-76177259 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esr2 | NM_010157.3 | 5 | 1100 | NP_034287.3 |
| NM_207707.1 | 5 | 1100 | NP_997590.1 | |
| NR_104386.1 | 5 | 1100 | ||
| AK054290.1 | 5 | 1098 | BAC35719.1 | |
| AK054301.1 | 5 | 1101 | ||
| AK054413.1 | 5 | 1101 | BAC35770.1 | |
| AY054413.1 | 4 | 561 | AAL15175.1 | |
| BC141075.1 | 4 | 802 | ||
| BC145329.1 | 4 | 802 | ||
| U81451.1 | 4 | 572 | AAB51132.1 |

Genomic Map