Assay Details
Target Gene Details
Entrez Gene ID: | 26380 |
Gene Name: | estrogen related receptor, beta |
Gene Aliases: |
Err2, Errb, Estrrb, Nr3b2 |
Location: |
Chr.12:86361117-86521628 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrrb | NM_001159500.1 | NP_001152972.1 | ||
| NM_011934.4 | 1 | 268 | NP_036064.3 | |
| AK032025.1 | 1 | 76 | BAC27656.1 | |
| AK044339.1 | 1 | 269 | ||
| AK052256.1 | BAC34898.1 | |||
| BB642270.1 | 1 | 269 | ||
| BC132595.1 | ||||
| BC132597.1 |

Genomic Map