Assay Details
Target Gene Details
Entrez Gene ID: | 26380 |
Gene Name: | estrogen related receptor, beta |
Gene Aliases: |
Err2, Errb, Estrrb, Nr3b2 |
Location: |
Chr.12:86361117-86521628 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrrb | NM_001159500.1 | 6 | 1275 | NP_001152972.1 |
| NM_011934.4 | 6 | 1174 | NP_036064.3 | |
| AK032025.1 | 6 | 982 | BAC27656.1 | |
| AK044339.1 | 6 | 1175 | ||
| AK052256.1 | 6 | 1277 | BAC34898.1 | |
| BC044858.1 | 5 | 844 | AAH44858.1 | |
| BC132595.1 | 6 | 1139 | ||
| BC132597.1 | 6 | 1140 | ||
| S82458.1 | 5 | 882 | AAB37687.1 | |
| X89594.1 | 5 | 882 | CAA61755.1 |

Genomic Map