Assay Details
Target Gene Details
Entrez Gene ID: | 20666 |
Gene Name: | SRY (sex determining region Y)-box 11 |
Gene Aliases: |
1110038H03Rik, 6230403H02Rik, AI836553, end1 |
Location: |
Chr.12:27334268-27342718 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox11 | NM_009234.6 | 1 | 7040 | NP_033260.4 |
| AK012306.1 | 1 | 733 |
More Information
Set Membership: |
|

Genomic Map