Assay Details
Target Gene Details
Entrez Gene ID: | 20677 |
Gene Name: | SRY (sex determining region Y)-box 4 |
Gene Aliases: |
AA682046, Sox-4 |
Location: |
Chr.13:28950716-28953682 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox4 | NM_009238.2 | 1 | 2469 | NP_033264.2 |
| AK205118.1 | 1 | 46 | ||
| BC052736.1 | 1 | 2469 | AAH52736.1 |
More Information
Set Membership: |
|

Genomic Map