Assay Details
Target Gene Details
Entrez Gene ID: | 70059 |
Gene Name: | degenerative spermatocyte homolog 2 (Drosophila), lipid desaturase |
Gene Aliases: |
2210008A03Rik, AI852933, DES2 |
Location: |
Chr.12:108686792-108702306 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Degs2 | NM_001171002.1 | 3 | 1863 | NP_001164473.1 |
Target Gene Details
Entrez Gene ID: | 14026 |
Gene Name: | Ena-vasodilator stimulated phosphoprotein |
Gene Aliases: |
AI528774 |
Location: |
Chr.12:108554720-108688516 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Evl | NM_001163394.1 | 14 | 1782 | NP_001156866.1 |
| NM_001163395.1 | 13 | 1471 | NP_001156867.1 | |
| NM_001163396.2 | 15 | 1556 | NP_001156868.1 | |
| NM_007965.3 | 13 | 1719 | NP_031991.3 | |
| AK134563.1 | 13 | 1472 | ||
| AK151633.1 | 16 | 1558 | ||
| BC059810.1 | 13 | 1719 | AAH59810.1 | |
| U72519.1 | 13 | 1482 | AAC52862.1 |
More Information
Set Membership: |
|

Genomic Map