Assay Details
Target Gene Details
Entrez Gene ID: | 80913 |
Gene Name: | pumilio RNA-binding family member 2 |
Gene Aliases: |
5730503J23Rik, Pumm2, mKIAA0235 |
Location: |
Chr.12:8674134-8752583 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pum2 | NM_001160219.1 | 21 | 4305 | NP_001153691.1 |
| NM_001160220.1 | 21 | 4299 | NP_001153692.1 | |
| NM_001160221.1 | 21 | 4114 | NP_001153693.1 | |
| NM_001160222.1 | 20 | 3877 | NP_001153694.1 | |
| NM_030723.2 | 21 | 4229 | NP_109648.2 | |
| NR_027670.1 | 22 | 4261 | ||
| AK134185.1 | 21 | 4113 | ||
| AK141682.1 | 21 | 4161 | BAE24797.1 | |
| AK147617.1 | 22 | 4247 | ||
| BC013765.1 | 5 | 1480 | AAH13765.1 |
More Information
Set Membership: |
|

Genomic Map