Assay Details
Target Gene Details
Entrez Gene ID: | 110095 |
Gene Name: | liver glycogen phosphorylase |
Gene Aliases: |
- |
Location: |
Chr.12:70190815-70227683 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pygl | NM_133198.2 | 1 | 158 | NP_573461.2 |
| AF288783.1 | 1 | 100 | AAG00588.1 | |
| AK041189.1 | 1 | 105 | BAC30855.1 | |
| AK140321.1 | 1 | 105 | BAE24332.1 | |
| AK145989.1 | 1 | 159 | BAE26811.1 | |
| AK167337.1 | 1 | 105 | BAE39438.1 | |
| BC013636.1 | 1 | 92 | AAH13636.1 |
More Information
Set Membership: |
|

Genomic Map