Assay Details
Target Gene Details
Entrez Gene ID: | 19325 |
Gene Name: | RAB10, member RAS oncogene family |
Gene Aliases: |
AW107754 |
Location: |
Chr.12:3247428-3309969 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab10 | NM_016676.5 | 6 | 2297 | NP_057885.1 |
| AK028320.1 | 6 | 2268 | BAC25878.1 | |
| AK087964.1 | 6 | 2259 | BAC40062.1 | |
| AK134988.1 | 6 | 2217 | BAE22371.1 | |
| AK146357.1 | 6 | 2298 | BAE27109.1 | |
| AK150319.1 | 6 | 2257 | BAE29464.1 | |
| AK150384.1 | 6 | 1951 | BAE29515.1 | |
| AK150791.1 | 6 | 2247 | BAE29855.1 | |
| AK151473.1 | 6 | 1951 | BAE30429.1 | |
| AK153315.1 | 6 | 2257 | BAE31896.1 | |
| AK153325.1 | 6 | 2247 | BAE31904.1 | |
| AK159383.1 | 6 | 2262 | BAE35038.1 | |
| AK159845.1 | 6 | 2262 | BAE35423.1 | |
| AK159988.1 | 6 | 2256 | BAE35538.1 | |
| AK160960.1 | 6 | 2252 | BAE36116.1 | |
| AK166010.1 | 6 | 2271 | ||
| AK168320.1 | 6 | 2257 | BAE40259.1 | |
| AK194711.1 | 1 | 168 | ||
| AK208805.1 | 1 | 203 | ||
| BC056374.1 | 6 | 2248 | AAH56374.1 |
Target Gene Details
Entrez Gene ID: | 74173 |
Gene Name: | RAB10, member RAS oncogene family, opposite strand |
Gene Aliases: |
1700012B15Rik, 1810036A22Rik |
Location: |
Chr.12:3235791-3250374 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab10os | NR_015551.1 | |||
| AK005738.1 | BAC25123.1 | |||
| AK007702.1 |
More Information
Set Membership: |
|

Genomic Map