Assay Details
Target Gene Details
Entrez Gene ID: | 52708 |
Gene Name: | zinc finger protein 410 |
Gene Aliases: |
Apa1, D12Ertd748e, Znf410 |
Location: |
Chr.12:84315928-84344439 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp410 | NM_144833.3 | 12 | 2226 | NP_659082.1 |
| NR_045546.1 | 12 | 2194 | ||
| NR_045615.1 | 10 | 1910 | ||
| AF295806.1 | 12 | 1988 | AAL82191.1 | |
| AK051939.1 | 10 | 1626 | ||
| AK083003.1 | 12 | 1969 | ||
| AK132345.1 | 12 | 1965 | ||
| BC021528.1 | 12 | 1981 | AAH21528.1 |
More Information
Set Membership: |
|

Genomic Map