Assay Details
Target Gene Details
Entrez Gene ID: | 218454 |
Gene Name: | lipoma HMGIC fusion partner-like 2 |
Gene Aliases: |
6030465B15, AI447312, AW050335, mKIAA0206, vgim |
Location: |
Chr.13:94057656-94195410 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhfpl2 | NM_172589.2 | 4 | 3158 | NP_766177.1 |
| AK030103.1 | 3 | 3316 | BAC26786.1 | |
| AK031635.1 | 4 | 3156 | ||
| AK052969.1 | 2 | 3020 | BAC35225.1 | |
| AK087506.1 | 4 | 3159 | BAC39903.1 | |
| AK215900.1 | ||||
| BC040389.1 | 2 | 2942 | AAH40389.1 |

Genomic Map