Assay Details
Target Gene Details
Entrez Gene ID: | 107513 |
Gene Name: | signal sequence receptor, alpha |
Gene Aliases: |
2510001K09Rik, 6330400D04, AI159733, AI452176, SSR, TRAPA |
Location: |
Chr.13:37971401-37994203 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssr1 | NM_025965.3 | 8 | 5111 | NP_080241.3 |

Genomic Map