Assay Details
Target Gene Details
Entrez Gene ID: | 20963 |
Gene Name: | spleen tyrosine kinase |
Gene Aliases: |
Sykb |
Location: |
Chr.13:52582329-52648792 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syk | NM_001198977.1 | 14 | 4072 | NP_001185906.1 |
| NM_011518.2 | 14 | 4076 | NP_035648.2 | |
| AK143564.1 | 14 | 4105 | BAE25438.1 | |
| BC065121.1 | 14 | 4008 | AAH65121.1 | |
| Z49877.1 | 14 | 4334 | CAA90034.1 |

Genomic Map