Assay Details
Target Gene Details
Entrez Gene ID: | 20963 |
Gene Name: | spleen tyrosine kinase |
Gene Aliases: |
Sykb |
Location: |
Chr.13:52582329-52648792 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syk | NM_001198977.1 | 2 | 325 | NP_001185906.1 |
| NM_011518.2 | 2 | 329 | NP_035648.2 | |
| AK036736.1 | 2 | 253 | BAE20505.1 | |
| AK049207.1 | 3 | 423 | ||
| AK136363.1 | 2 | 328 | BAE22946.1 | |
| AK138741.1 | 2 | 260 | ||
| AK143564.1 | 2 | 358 | BAE25438.1 | |
| BB629720.1 | 2 | 251 | ||
| BC065121.1 | 2 | 261 | AAH65121.1 | |
| U25685.1 | 1 | 113 | AAA87462.1 | |
| U36776.1 | 2 | 330 | AAA79996.1 | |
| Z49877.1 | 2 | 590 | CAA90034.1 |
More Information
Set Membership: |
|

Genomic Map