Assay Details
Target Gene Details
Entrez Gene ID: | 238683 |
Gene Name: | spermatogenesis associated 31 subfamily D, member 1C |
Gene Aliases: |
4932441B19Rik, Fam75d1c |
Location: |
Chr.13:65033058-65038004 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata31d1c | NM_001083890.2 | 4 | 1334 | NP_001077359.2 |
| AK133350.1 | 4 | 1335 | BAE21611.1 |

Genomic Map