Assay Details
Target Gene Details
Entrez Gene ID: | 12061 |
Gene Name: | bradykinin receptor, beta 1 |
Gene Aliases: |
B1BKR, B1R, BKR1, BRADYB1, Bdkrb |
Location: |
Chr.12:105604091-105605428 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdkrb1 | NM_007539.2 | 1 | 448 | NP_031565.1 |
| BC120684.1 | 1 | 410 |
More Information
Set Membership: |
|

Genomic Map