Assay Details
Target Gene Details
Entrez Gene ID: | 104943 |
Gene Name: | family with sequence similarity 110, member C |
Gene Aliases: |
9030611O19Rik, AU019489 |
Location: |
Chr.12:31073968-31079940 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam110c | NM_027828.2 | 2 | 2004 | NP_082104.2 |
| AK139763.1 | 2 | 2113 | ||
| AK160079.1 | 2 | 2008 | BAE35611.1 | |
| BC019466.1 | 2 | 2004 | AAH19466.1 | |
| BC085288.1 | 2 | 1989 | AAH85288.1 |

Genomic Map