Assay Details
Target Gene Details
Entrez Gene ID: | 217705 |
Gene Name: | family with sequence similarity 161, member B |
Gene Aliases: |
9330169D17, 9830169C18Rik |
Location: |
Chr.12:84345314-84361883 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam161b | NM_172581.2 | 4 | 1231 | NP_766169.1 |
| AK031252.1 | 4 | 1182 | BAC27322.1 | |
| AK034254.1 | 4 | 1234 | BAC28650.1 | |
| AK036740.1 | 4 | 1232 | BAC29559.1 | |
| AK046195.1 | 3 | 2253 | BAC32631.1 | |
| BC062132.1 | 4 | 1224 | AAH62132.1 |

Genomic Map