Assay Details
Target Gene Details
Entrez Gene ID: | 72350 |
Gene Name: | zinc finger, C2HC-type containing 1C |
Gene Aliases: |
2810002I04Rik, AV046379, Fam164c |
Location: |
Chr.12:85274388-85299361 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc2hc1c | NM_172414.4 | 2 | 422 | NP_766002.2 |
| AK012642.1 | 2 | 307 | ||
| AK032455.1 | 2 | 326 | BAC27877.1 | |
| AK033213.1 | 2 | 317 | BAC28200.1 | |
| AK036602.1 | 2 | 360 | BAC29503.1 | |
| BC049940.1 | 2 | 336 | AAH49940.1 | |
| BX525619.1 | 2 | 447 |

Genomic Map