Assay Details
Target Gene Details
Entrez Gene ID: | 72350 |
Gene Name: | zinc finger, C2HC-type containing 1C |
Gene Aliases: |
2810002I04Rik, AV046379, Fam164c |
Location: |
Chr.12:85274388-85299361 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc2hc1c | NM_172414.4 | 3 | 2507 | NP_766002.2 |
| AK032455.1 | BAC27877.1 | |||
| AK036602.1 | 3 | 2445 | BAC29503.1 |

Genomic Map