Assay Details
Target Gene Details
Entrez Gene ID: | 209550 |
Gene Name: | RAD51 associated protein 2 |
Gene Aliases: |
EG209550 |
Location: |
Chr.12:11455759-11517161 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad51ap2 | NM_001111118.1 | 1 | 454 | NP_001104588.1 |

Genomic Map