Assay Details
Target Gene Details
Entrez Gene ID: | 193385 |
Gene Name: | family with sequence similarity 65, member B |
Gene Aliases: |
- |
Location: |
Chr.13:24501534-24733806 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam65b | NM_001080381.1 | 15 | 2389 | NP_001073850.1 |
| NM_001286100.1 | NP_001273029.1 | |||
| NM_001286101.1 | 15 | 2356 | NP_001273030.1 | |
| NM_029679.2 | NP_083955.1 | |||
| NM_178658.5 | 14 | 2116 | NP_848773.3 | |
| AK031928.1 | 15 | 2342 | BAC27606.1 | |
| AK036452.1 | 15 | 2336 | BAC29436.1 | |
| AK036695.1 | 3 | 1006 | ||
| AK040887.1 | 14 | 2096 | BAC30732.1 | |
| AK122269.1 | BAC65551.1 | |||
| AK134538.1 |
More Information
Set Membership: |
|

Genomic Map