Assay Details
Target Gene Details
Entrez Gene ID: | 17084 |
Gene Name: | lymphocyte antigen 86 |
Gene Aliases: |
MD-1, MD1 |
Location: |
Chr.13:37345345-37419036 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ly86 | NM_010745.2 | 5 | 568 | NP_034875.1 |
| AB007599.1 | 5 | 568 | BAA32399.1 | |
| AK027929.1 | 5 | 557 | BAC25670.1 | |
| BC065783.1 | 5 | 473 | AAH65783.1 | |
| BC096414.1 | 5 | 536 | AAH96414.1 | |
| BE200280.1 | 1 | 430 |

Genomic Map