Assay Details
Target Gene Details
Entrez Gene ID: | 105504 |
Gene Name: | exocyst complex component 5 |
Gene Aliases: |
AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1 |
Location: |
Chr.14:49011922-49066667 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exoc5 | NM_207214.3 | 18 | 3194 | NP_997097.1 |
| AK134562.1 | 18 | 2707 | ||
| AK150297.1 | 7 | 1461 | BAE29449.1 | |
| AK157473.1 | 18 | 2889 | BAE34093.1 | |
| AK170148.1 | 18 | 2888 |
More Information
Set Membership: |
|

Genomic Map