Assay Details
Target Gene Details
Entrez Gene ID: | 219114 |
Gene Name: | spindle and kinetochore associated complex subunit 3 |
Gene Aliases: |
F630043A04Rik |
Location: |
Chr.14:57806560-57826190 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ska3 | NM_198605.3 | 4 | 524 | NP_941007.1 |
| AK089223.1 | 4 | 525 | BAC40798.1 | |
| AK158142.1 | 4 | 546 | ||
| AK166171.1 | 4 | 515 | ||
| BC117500.1 | 4 | 516 | ||
| BC117501.1 | 4 | 516 |

Genomic Map