Assay Details
Target Gene Details
Entrez Gene ID: | 12763 |
Gene Name: | cytidine monophospho-N-acetylneuraminic acid hydroxylase |
Gene Aliases: |
- |
Location: |
Chr.13:24326980-24477289 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmah | NM_001111110.2 | 2 | 183 | NP_001104580.1 |
| NM_001284519.1 | 3 | 345 | NP_001271448.1 | |
| AB061276.1 | 2 | 167 | BAB91361.1 | |
| AB061277.1 | 3 | 329 | BAB91362.1 |
More Information
Set Membership: |
|

Genomic Map